Assay Details
| Gene Symbol: Sytbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140477.2 | NP_648734.1 | 6 - 7 | 1882 | 87 | |
| NM_144170.2 | NP_652427.2 | 6 - 7 | 1784 | 87 | ||
| GenBank mRNA | BT133101.1 | - | 6 - 7 | 1785 | 87 | |
More Information
Set Membership: |
|

Sytbeta
Genomic Map