Assay Details
| Gene Symbol: CG17028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202185.1 | NP_001189114.1 | 1 - 2 | 497 | 87 | |
| NM_140565.2 | NP_648822.1 | 1 - 2 | 497 | 87 | ||
More Information
Set Membership: |
|

CG17028
Genomic Map