Assay Details
| Gene Symbol: CG17027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274980.1 | NP_001261909.1 | 1 - 2 | 849 | 81 | |
| NM_140566.3 | NP_648823.1 | 1 - 2 | 653 | 81 | ||
| GenBank mRNA | AY069101.1 | - | 1 - 2 | 653 | 81 | |
More Information
Set Membership: |
|

CG17027
Genomic Map