Assay Details
| Gene Symbol: ClC-c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274993.1 | NP_001261922.1 | 7 - 8 | 2595 | 67 | |
| NM_140577.2 | NP_648834.1 | 7 - 8 | 2196 | 67 | ||
| NM_168650.2 | NP_730105.3 | 7 - 8 | 2595 | 67 | ||
| GenBank mRNA | BT133258.1 | - | 7 - 8 | 2595 | 67 | |
More Information
Set Membership: |
|

ClC-c
Genomic Map