Assay Details
| Gene Symbol: Cpr72Eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140626.2 | NP_648883.1 | 1 | 711 | 69 | |
| GenBank mRNA | BT100091.4 | - | 1 | 613 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr72Eb| Gene Symbol: Cpr72Eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140626.2 | NP_648883.1 | 1 | 711 | 69 | |
| GenBank mRNA | BT100091.4 | - | 1 | 613 | 69 | |
Set Membership: |
|