Assay Details
| Gene Symbol: RAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275014.1 | NP_001261943.1 | 3 - 4 | 2898 | 69 | |
| NM_140632.3 | NP_648889.2 | 3 - 4 | 2828 | 69 | ||
| NM_168664.2 | NP_730163.1 | 3 - 4 | 2904 | 69 | ||
| GenBank mRNA | BT021383.1 | - | 3 - 4 | 2886 | 69 | |
More Information
Set Membership: |
|

RAF2
Genomic Map