Assay Details
| Gene Symbol: Gagr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300152.1 | NP_001287081.1 | 1 | 1680 | 73 | |
| NM_140634.2 | NP_648891.1 | 1 | 1428 | 73 | ||
| GenBank mRNA | AY047572.1 | - | 1 | 1418 | 73 | |
More Information
Set Membership: |
|

Gagr
Genomic Map