Assay Details
| Gene Symbol: TyrRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140638.3 | NP_648895.1 | 1 - 2 | 703 | 82 | |
| GenBank mRNA | AY051662.1 | - | 1 - 2 | 696 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TyrRS| Gene Symbol: TyrRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140638.3 | NP_648895.1 | 1 - 2 | 703 | 82 | |
| GenBank mRNA | AY051662.1 | - | 1 - 2 | 696 | 82 | |
Set Membership: |
|