Assay Details
| Gene Symbol: TyrRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140638.3 | NP_648895.1 | 2 - 3 | 1068 | 67 | |
| GenBank mRNA | AY051662.1 | - | 2 - 3 | 1061 | 67 | |
More Information
Set Membership: |
|

TyrRS
Genomic Map