Assay Details
| Gene Symbol: CG42514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140642.3 | NP_648899.2 | 2 - 3 | 1153 | 70 | |
| NM_168668.2 | NP_730171.1 | 3 - 4 | 1216 | 70 | ||
| NM_168669.2 | NP_730172.1 | 3 - 4 | 1130 | 70 | ||
| GenBank mRNA | BT100042.1 | - | 3 - 4 | 1111 | 70 | |
More Information
Set Membership: |
|

CG42514
Genomic Map