Assay Details
| Gene Symbol: CG9674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259874.1 | NP_001246803.1 | 12 - 13 | 4514 | 81 | |
| NM_001259875.2 | NP_001246804.1 | 11 - 12 | 4021 | 81 | ||
| NM_001275044.1 | NP_001261973.1 | 11 - 12 | 3992 | 81 | ||
| NM_001275045.1 | NP_001261974.1 | 12 - 13 | 4506 | 81 | ||
| NM_001300159.1 | NP_001287088.1 | 10 - 11 | 4739 | 81 | ||
| NM_140665.2 | NP_648922.1 | 11 - 12 | 3989 | 81 | ||
| NM_176339.2 | NP_788517.1 | 11 - 12 | 4099 | 81 | ||
| GenBank mRNA | AY094734.1 | - | 11 - 12 | 3951 | 81 | |
More Information
Set Membership: |
|

CG9674
Genomic Map