Assay Details
| Gene Symbol: U4-U6-60K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140733.3 | NP_648990.1 | 1 | 1853 | 155 | |
| GenBank mRNA | AY061623.1 | - | 1 | 1814 | 168 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

U4-U6-60K| Gene Symbol: U4-U6-60K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140733.3 | NP_648990.1 | 1 | 1853 | 155 | |
| GenBank mRNA | AY061623.1 | - | 1 | 1814 | 168 | |
Set Membership: |
|