Assay Details
| Gene Symbol: MED19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300174.1 | NP_001287103.1 | 1 - 2 | 748 | 72 | |
| NM_140759.3 | NP_649016.1 | 1 - 2 | 748 | 72 | ||
| GenBank mRNA | AY069647.1 | - | 1 - 2 | 729 | 72 | |
More Information
Set Membership: |
|

MED19
Genomic Map