Assay Details
| Gene Symbol: AstC-R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140783.2 | NP_649040.2 | 3 - 4 | 1490 | 62 | |
| GenBank mRNA | AY061635.1 | - | 3 - 4 | 1490 | 62 | |
More Information
Set Membership: |
|

AstC-R1
Genomic Map