Assay Details
| Gene Symbol: CG18234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140786.2 | NP_649043.3 | 1 - 2 | 198 | 78 | |
| GenBank mRNA | BT022668.1 | - | 1 - 2 | 185 | 78 | |
More Information
Set Membership: |
|

CG18234
Genomic Map