Assay Details
| Gene Symbol: CG6841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316442.1 | NP_001303371.1 | 1 - 2 | 220 | 73 | |
| NM_140816.3 | NP_649073.1 | 1 - 2 | 220 | 73 | ||
| GenBank mRNA | BT021415.1 | - | 1 - 2 | 180 | 73 | |
More Information
Set Membership: |
|

CG6841
Genomic Map