Assay Details
| Gene Symbol: CG6841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316442.1 | NP_001303371.1 | 3 - 4 | 2692 | 64 | |
| NM_140816.3 | NP_649073.1 | 3 - 4 | 2692 | 64 | ||
| GenBank mRNA | BT021415.1 | - | 3 - 4 | 2652 | 64 | |
More Information
Set Membership: |
|

CG6841
Genomic Map