Assay Details
| Gene Symbol: Gem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275105.1 | NP_001262034.1 | 2 - 3 | 247 | 103 | |
| NM_140835.4 | NP_649092.1 | 1 - 2 | 306 | 103 | ||
More Information
Set Membership: |
|

Gem2
Genomic Map