Assay Details
| Gene Symbol: Cpr76Ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140863.2 | NP_649120.1 | 1 | 631 | 65 | |
| GenBank mRNA | BT100095.4 | - | 1 | 574 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr76Ba| Gene Symbol: Cpr76Ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140863.2 | NP_649120.1 | 1 | 631 | 65 | |
| GenBank mRNA | BT100095.4 | - | 1 | 574 | 65 | |
Set Membership: |
|