Assay Details
| Gene Symbol: l(3)76BDm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275129.1 | NP_001262058.1 | 3 - 4 | 1330 | 63 | |
| NM_140878.4 | NP_649135.2 | 3 - 4 | 1330 | 63 | ||
| GenBank mRNA | BT133116.1 | - | 3 - 4 | 1326 | 63 | |
More Information
Set Membership: |
|

l(3)76BDm
Genomic Map