Assay Details
| Gene Symbol: Mi-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014591.3 | NP_001014591.1 | 3 - 4 | 499 | 59 | |
| NM_001170005.2 | NP_001163476.1 | 3 - 4 | 496 | 59 | ||
| NM_001275149.1 | NP_001262078.1 | 3 - 4 | 437 | 59 | ||
| NM_140897.4 | NP_649154.2 | 3 - 4 | 496 | 59 | ||
More Information
Set Membership: |
|

Mi-2
Genomic Map