Assay Details
| Gene Symbol: Mi-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140897.4 | NP_649154.2 | 5 - 6 | 6163 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mi-2| Gene Symbol: Mi-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140897.4 | NP_649154.2 | 5 - 6 | 6163 | 125 | |
Set Membership: |
|