Assay Details
| Gene Symbol: Toll-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259916.2 | NP_001246845.1 | 3 - 4 | 2148 | 80 | |
| NM_001259917.1 | NP_001246846.1 | 4 - 5 | 2370 | 80 | ||
| NM_140957.4 | NP_649214.1 | 5 - 6 | 2955 | 80 | ||
More Information
Set Membership: |
|

Toll-9
Genomic Map