Assay Details
| Gene Symbol: CG13248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140962.2 | NP_649219.1 | 1 - 2 | 305 | 57 | |
| GenBank mRNA | BT099767.1 | - | 1 - 2 | 304 | 57 | |
More Information
Set Membership: |
|

CG13248
Genomic Map