Assay Details
| Gene Symbol: CtIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140967.3 | NP_649224.1 | 2 - 3 | 806 | 66 | |
| GenBank mRNA | AY071347.1 | - | 2 - 3 | 799 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CtIP| Gene Symbol: CtIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140967.3 | NP_649224.1 | 2 - 3 | 806 | 66 | |
| GenBank mRNA | AY071347.1 | - | 2 - 3 | 799 | 66 | |
Set Membership: |
|