Assay Details
| Gene Symbol: l(3)77CDf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275189.1 | NP_001262118.1 | 1 - 2 | 685 | 64 | |
| NM_140985.4 | NP_649242.1 | 1 - 2 | 685 | 64 | ||
| GenBank mRNA | AY069402.1 | - | 1 - 2 | 692 | 64 | |
More Information
Set Membership: |
|

l(3)77CDf
Genomic Map