Assay Details
| Gene Symbol: l(3)77CDf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140985.4 | NP_649242.1 | 4 - 5 | 1728 | 63 | |
| GenBank mRNA | AY069402.1 | - | 4 - 5 | 1735 | 63 | |
More Information
Set Membership: |
|

l(3)77CDf
Genomic Map