Assay Details
| Gene Symbol: CG11396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140998.3 | NP_649255.1 | 1 - 2 | 351 | 50 | |
| GenBank mRNA | AY089596.1 | - | 1 - 2 | 348 | 50 | |
More Information
Set Membership: |
|

CG11396
Genomic Map