Assay Details
| Gene Symbol: CG44085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169497.1 | NP_001162968.1 | 2 - 3 | 737 | 96 | |
| NM_001169498.1 | NP_001162969.1 | 2 - 3 | 502 | 96 | ||
| NM_001169499.1 | NP_001162970.1 | 2 - 3 | 494 | 96 | ||
| NM_001259090.1 | NP_001246019.1 | 1 - 2 | 477 | 96 | ||
| NM_001259091.2 | NP_001246020.1 | 1 - 2 | 494 | 96 | ||
| NM_001273511.1 | NP_001260440.1 | 1 - 2 | 494 | 96 | ||
| NM_165034.3 | NP_723801.2 | 1 - 2 | 494 | 96 | ||
| NM_165037.3 | NP_723804.1 | 3 - 4 | 736 | 96 | ||
| GenBank mRNA | BT083391.1 | - | 2 - 3 | 728 | 96 | |
| BT088972.1 | - | 1 - 2 | 477 | 96 | ||
More Information
Set Membership: |
|

CG44085
Genomic Map