Assay Details
| Gene Symbol: kuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014481.3 | NP_001014481.1 | 4 - 5 | 910 | 141 | |
| NM_001103689.2 | NP_001097159.2 | 4 - 5 | 1037 | 141 | ||
| NM_001103690.3 | NP_001097160.2 | 4 - 5 | 910 | 141 | ||
| NM_057839.5 | NP_477187.1 | 4 - 5 | 1037 | 141 | ||
| NM_165047.3 | NP_723825.1 | 4 - 5 | 910 | 141 | ||
| GenBank mRNA | BT150472.1 | - | 4 - 5 | 891 | 141 | |
More Information
Set Membership: |
|

kuz
Genomic Map