Assay Details
| Gene Symbol: Ance | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298986.1 | NP_001285915.1 | 6 - 7 | 1935 | 109 | |
| NM_057698.5 | NP_477046.1 | 6 - 7 | 1933 | 109 | ||
| NM_165070.3 | NP_723852.1 | 5 - 6 | 1991 | 109 | ||
| GenBank mRNA | AY061129.1 | - | 6 - 7 | 1912 | 109 | |
More Information
Set Membership: |
|

Ance
Genomic Map