Assay Details
| Gene Symbol: NimB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165075.3 | NP_723857.1 | 2 - 3 | 1053 | 62 | |
| GenBank mRNA | BT012496.1 | - | 2 - 3 | 1042 | 62 | |
More Information
Set Membership: |
|

NimB2
Genomic Map