Assay Details
| Gene Symbol: wb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201877.2 | NP_001188806.1 | 5 - 6 | 3791 | 68 | |
| NM_001201878.2 | NP_001188807.1 | 5 - 6 | 3873 | 68 | ||
| NM_001201879.2 | NP_001188808.1 | 5 - 6 | 3999 | 68 | ||
| NM_001201880.2 | NP_001188809.1 | 5 - 6 | 3812 | 68 | ||
| NM_057442.5 | NP_476790.3 | 7 - 8 | 6149 | 68 | ||
| NM_165086.3 | NP_723870.1 | 7 - 8 | 6149 | 68 | ||
More Information
Set Membership: |
|

wb
Genomic Map