Assay Details
| Gene Symbol: vas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273529.2 | NP_001260458.1 | 3 - 4 | 568 | 62 | |
| NM_001316393.1 | NP_001303322.1 | 3 - 4 | 522 | 62 | ||
| NM_165103.3 | NP_723899.1 | 3 - 4 | 568 | 62 | ||
More Information
Set Membership: |
|

vas
Genomic Map