Assay Details
| Gene Symbol: BicC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057517.3 | NP_476865.1 | 3 - 4 | 1193 | 115 | |
| NM_165144.2 | NP_723948.1 | 2 - 3 | 870 | 115 | ||
| GenBank mRNA | BT023837.1 | - | 3 - 4 | 1110 | 115 | |
| Gene Symbol: CG45084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299044.2 | NP_001285973.1 | 2 - 3 | 870 | 115 | |
| NM_001299045.1 | NP_001285974.1 | 3 - 4 | 1193 | 115 | ||
More Information
Set Membership: |
|

BicC
Genomic Map