Assay Details
| Gene Symbol: BicC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057517.3 | NP_476865.1 | 10 - 11 | 3345 | 72 | |
| NM_165144.2 | NP_723948.1 | 9 - 10 | 3022 | 72 | ||
| NM_165145.1 | NP_723949.1 | 6 - 7 | 2341 | 72 | ||
| GenBank mRNA | BT023837.1 | - | 10 - 11 | 3262 | 72 | |
| Gene Symbol: CG45084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299044.2 | NP_001285973.1 | 9 - 10 | 3022 | 72 | |
| NM_001299045.1 | NP_001285974.1 | 10 - 11 | 3345 | 72 | ||
More Information
Set Membership: |
|

BicC
Genomic Map