Assay Details
| Gene Symbol: Ca-alpha1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273551.1 | NP_001260480.2 | 15 - 16 | 4909 | 74 | |
| NM_001273552.1 | NP_001260481.1 | 14 - 15 | 3816 | 74 | ||
| NM_001273553.1 | NP_001260482.1 | 14 - 15 | 3816 | 74 | ||
| NM_001273554.1 | NP_001260483.1 | 14 - 15 | 3816 | 74 | ||
| NM_001273555.1 | NP_001260484.1 | 14 - 15 | 3816 | 74 | ||
| NM_001273556.1 | NP_001260485.1 | 14 - 15 | 3816 | 74 | ||
| NM_080365.3 | NP_525104.3 | 15 - 16 | 4899 | 74 | ||
| NM_134429.3 | NP_602305.2 | 15 - 16 | 4899 | 74 | ||
| NM_165146.2 | NP_723952.2 | 15 - 16 | 4813 | 74 | ||
| NM_165147.2 | NP_723953.2 | 15 - 16 | 4813 | 74 | ||
More Information
Set Membership: |
|

Ca-alpha1D
Genomic Map