Assay Details
| Gene Symbol: Ca-alpha1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273552.1 | NP_001260481.1 | 15 - 16 | 3874 | 133 | |
| NM_001273553.1 | NP_001260482.1 | 15 - 16 | 3874 | 133 | ||
| NM_001273554.1 | NP_001260483.1 | 15 - 16 | 3874 | 133 | ||
| NM_001273556.1 | NP_001260485.1 | 15 - 16 | 3874 | 133 | ||
| NM_134429.3 | NP_602305.2 | 16 - 17 | 4957 | 133 | ||
| NM_165146.2 | NP_723952.2 | 16 - 17 | 4871 | 133 | ||
| NM_165147.2 | NP_723953.2 | 16 - 17 | 4871 | 133 | ||
More Information
Set Membership: |
|

Ca-alpha1D
Genomic Map