Assay Details
| Gene Symbol: Ca-alpha1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273551.1 | NP_001260480.2 | 21 - 22 | 5577 | 126 | |
| NM_001273554.1 | NP_001260483.1 | 20 - 21 | 4490 | 126 | ||
| NM_001273556.1 | NP_001260485.1 | 21 - 22 | 4598 | 126 | ||
| NM_165146.2 | NP_723952.2 | 22 - 23 | 5595 | 126 | ||
More Information
Set Membership: |
|

Ca-alpha1D
Genomic Map