Assay Details
| Gene Symbol: cact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273567.1 | NP_001260496.1 | 2 - 3 | 910 | 75 | |
| NM_057594.4 | NP_476942.1 | 2 - 3 | 855 | 75 | ||
| NM_057595.4 | NP_476943.1 | 2 - 3 | 855 | 75 | ||
| NM_165152.2 | NP_723960.1 | 2 - 3 | 804 | 75 | ||
| NM_205999.2 | NP_995721.1 | 1 - 2 | 1089 | 75 | ||
| GenBank mRNA | AY069399.1 | - | 1 - 2 | 1112 | 75 | |
More Information
Set Membership: |
|

cact
Genomic Map