Assay Details
| Gene Symbol: cact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273567.1 | NP_001260496.1 | 5 - 6 | 1387 | 64 | |
| NM_057594.4 | NP_476942.1 | 5 - 6 | 1332 | 64 | ||
| NM_057595.4 | NP_476943.1 | 5 - 6 | 1332 | 64 | ||
| NM_165152.2 | NP_723960.1 | 5 - 6 | 1281 | 64 | ||
| NM_205999.2 | NP_995721.1 | 4 - 5 | 1566 | 64 | ||
| GenBank mRNA | AY069399.1 | - | 4 - 5 | 1589 | 64 | |
More Information
Set Membership: |
|

cact
Genomic Map