Assay Details
| Gene Symbol: Tpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273571.1 | NP_001260500.1 | 2 - 3 | 608 | 110 | |
| NM_001273572.1 | NP_001260501.1 | 2 - 3 | 963 | 110 | ||
| NM_001273573.1 | NP_001260502.1 | 3 - 4 | 1104 | 110 | ||
| NM_078860.3 | NP_523584.1 | 2 - 3 | 963 | 110 | ||
| NM_165164.2 | NP_723974.1 | 2 - 3 | 834 | 110 | ||
| GenBank mRNA | AF199022.1 | - | 2 - 3 | 956 | 110 | |
| BT100099.1 | - | 2 - 3 | 837 | 110 | ||
More Information
Set Membership: |
|

Tpr2
Genomic Map