Assay Details
| Gene Symbol: grp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057663.4 | NP_477011.1 | 2 - 3 | 846 | 60 | |
| NM_165171.3 | NP_723985.1 | 2 - 3 | 517 | 60 | ||
| NM_165172.3 | NP_723986.1 | 3 - 4 | 627 | 60 | ||
| GenBank mRNA | AY051961.1 | - | 2 - 3 | 821 | 60 | |
More Information
Set Membership: |
|

grp
Genomic Map