Assay Details
| Gene Symbol: grp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273579.1 | NP_001260508.1 | 5 - 6 | 1036 | 65 | |
| NM_057663.4 | NP_477011.1 | 6 - 7 | 1589 | 65 | ||
| NM_165171.3 | NP_723985.1 | 6 - 7 | 1260 | 65 | ||
| NM_165172.3 | NP_723986.1 | 7 - 8 | 1370 | 65 | ||
| NM_165173.3 | NP_723987.2 | 4 - 5 | 675 | 65 | ||
| GenBank mRNA | AY051961.1 | - | 6 - 7 | 1564 | 65 | |
More Information
Set Membership: |
|

grp
Genomic Map