Assay Details
| Gene Symbol: VhaSFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273581.1 | NP_001260510.1 | 3 - 4 | 811 | 58 | |
| NM_078861.3 | NP_523585.2 | 3 - 4 | 811 | 58 | ||
| NM_165177.2 | NP_723992.1 | 4 - 5 | 1033 | 58 | ||
| NM_165178.2 | NP_723993.1 | 3 - 4 | 811 | 58 | ||
| GenBank mRNA | AY052122.1 | - | 3 - 4 | 784 | 58 | |
More Information
Set Membership: |
|

VhaSFD
Genomic Map