Assay Details
| Gene Symbol: VhaSFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273581.1 | NP_001260510.1 | 5 - 6 | 1486 | 64 | |
| NM_078861.3 | NP_523585.2 | 5 - 6 | 1486 | 64 | ||
| NM_165177.2 | NP_723992.1 | 6 - 7 | 1708 | 64 | ||
| GenBank mRNA | AY052122.1 | - | 5 - 6 | 1459 | 64 | |
More Information
Set Membership: |
|

VhaSFD
Genomic Map