Assay Details
| Gene Symbol: Mhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165185.3 | NP_724003.1 | 10 - 11 | 2284 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mhc| Gene Symbol: Mhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165185.3 | NP_724003.1 | 10 - 11 | 2284 | 97 | |
Set Membership: |
|