Assay Details
| Gene Symbol: Mhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169521.2 | NP_001162992.1 | 10 - 11 | 2288 | 70 | |
| NM_001259119.2 | NP_001246048.1 | 10 - 11 | 2288 | 70 | ||
| NM_165189.3 | NP_724007.1 | 10 - 11 | 2288 | 70 | ||
| NM_165191.3 | NP_724009.1 | 10 - 11 | 2288 | 70 | ||
| NM_165192.4 | NP_724010.1 | 10 - 11 | 2288 | 70 | ||
More Information
Set Membership: |
|

Mhc
Genomic Map