Assay Details
| Gene Symbol: Mhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165190.4 | NP_724008.1 | 17 - 18 | 6095 | 136 | |
| NM_165191.3 | NP_724009.1 | 17 - 18 | 5927 | 136 | ||
| NM_165192.4 | NP_724010.1 | 17 - 18 | 5927 | 136 | ||
More Information
Set Membership: |
|

Mhc
Genomic Map