Assay Details
| Gene Symbol: AspRS-m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273590.1 | NP_001260519.1 | 3 - 4 | 360 | 90 | |
| NM_001273591.1 | NP_001260520.2 | 2 - 3 | 460 | 90 | ||
| NM_165196.2 | NP_724018.1 | 3 - 4 | 494 | 90 | ||
| GenBank mRNA | AY119180.1 | - | 3 - 4 | 474 | 90 | |
More Information
Set Membership: |
|

AspRS-m
Genomic Map